Learn More
Hampton Research CRYSTAL SCREEN NO04 185ML
SDP
Supplier:  Hampton Research HR291004
Encompass
HR2-910-04/Crystal Screen #04 (A4), 185 ml - 0.1 M TRIS hydrochloride pH 8.5, 2.0 M Ammonium sulfate. Please note items from this supplier are non-returnable.
Catalog No. 50-141-3