Learn More
Hampton Research CRYSTAL SCREEN NO05 185ML
SDP
Supplier:  Hampton Research HR291005
Encompass
HR2-910-05/Crystal Screen #05 (A5), 185 ml - 0.2 M Sodium citrate tribasic dihydrate, 0.1 M HEPES sodium pH 7.5, 30% v/v (+/-)-2-Methyl-2,4-pentanediol. Please note items from this supplier are non-returnable.
Catalog No. 50-141-4