Learn More
Hampton Research CRYSTAL SCREEN NO12 185ML
SDP
Supplier:  Hampton Research HR291012
Encompass
HR2-910-12/Crystal Screen #12 (A12), 185 ml - 0.2 M Magnesium chloride hexahydrate, 0.1 M HEPES sodium pH 7.5, 30% v/v 2-Propanol. Please note items from this supplier are non-returnable.
Catalog No. 50-142-1