Learn More
Hampton Research CRYSTAL SCREEN NO23 185ML
SDP
Supplier:  Hampton Research HR291023
Encompass
HR2-910-23/Crystal Screen #23 (B11), 185 ml - 0.2 M Magnesium chloride hexahydrate, 0.1 M HEPES sodium pH 7.5, 30% v/v Polyethylene glycol 400. Please note items from this supplier are non-returnable.
Catalog No. 50-143-2