Learn More
Hampton Research CRYSTAL SCREEN NO24 185ML
SDP
Supplier:  Hampton Research HR291024
Encompass
HR2-910-24/Crystal Screen #24 (B12), 185 ml - 0.2 M Calcium chloride dihydrate, 0.1 M Sodium acetate trihydrate pH 4.6, 20% v/v 2-Propanol. Please note items from this supplier are non-returnable.
Catalog No. 50-143-3