Learn More
Hampton Research CRYSTAL SCREEN NO27 185ML
SDP
Supplier:  Hampton Research HR291027
Encompass
HR2-910-27/Crystal Screen #27 (C3), 185 ml - 0.2 M Sodium citrate tribasic dihydrate, 0.1 M HEPES sodium pH 7.5, 20% v/v 2-Propanol. Please note items from this supplier are non-returnable.
Catalog No. 50-143-6