Learn More
Hampton Research CRYSTAL SCREEN NO41 185ML
SDP
Supplier:  Hampton Research HR291041
Encompass
HR2-910-41/Crystal Screen #41 (D5), 185 ml - 0.1 M HEPES sodium pH 7.5, 10% v/v 2-Propanol, 20% w/v Polyethylene glycol 4,000. Please note items from this supplier are non-returnable.
Catalog No. 50-145-0