Learn More
Volu Sol Crystal Violet Stain, 5 gal / 20 L, Volu-Sol™
SDP
Supplier:  Volu Sol VGS640
Encompass_Preferred
Crystal Violet, component of the Gram Stain Kit.  This solution acts as the primary stain in Gram procedures
Catalog No. 50-420-799