Learn More
Abcam Crystal violet, Cation-based violet dye, 25G
Supplier:  Abcam AB14309525G
Encompass_Preferred
MW 408 Da. Cation-based violet dye. Interacts with aqueous KI - I₂ during Gram staining to produce a precipitate. Histological stain for Gram-positive bacteria and DNA. Shows antibacterial, antifungal, and anthelmintic effects in vivo. .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6135