Learn More
Medchemexpress LLC Crystal violet IND 50g
SDP
Supplier:  Medchemexpress LLC HYW11079450G
Encompass_Preferred
Crystal violet IND (0 0-2 0) is a specialized indicator suitable for strongly acidic systems which reflects changes in pH within the range of 0 0-2 0 through color changes Crystal violet IND (0 0-2 0) is widely used in acid-base titration chemical analysis and industrial testing fields[1]
Catalog No. 50-002-06344