Learn More
Medchemexpress LLC CS-VIP 8 hydrochloride | 961.40 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYP10948A10MG
Encompass_Preferred
CS-VIP 8 hydrochloride is a selective allosteric WDR5 protein inhibitor (Ki = 0.008 μM). It induces conformational changes in the MLL1 complex, leading to the dissociation of MLL1 from the complex, inhibiting MLL1 histone methyltransferase activity, and regulating HOX gene expression.
  • Promising for research of hematological diseases such as leukemia
Catalog No. 50-004-22420