Learn More
Selleck Chemical LLC BLZ945 5mg 953769-46-5
SDP
Supplier:  Selleck Chemical LLC S77255MG
Encompass_Preferred
BLZ945 is an orally active, potent and selective CSF-1R inhibitor with IC50 of 1 nM, >1000-fold selective against its closest receptor tyrosine kinase homologs. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1276953