Learn More
Medchemexpress LLC CSF1R Mouse HEK293 50ug
SDP
Supplier:  Medchemexpress LLC HYP7295150UG
Encompass_Preferred
The CSF1R protein is a tyrosine protein kinase receptor for CSF1 and IL34 and plays a critical regulatory role in hematopoietic cells especially mononuclear phagocytes Its effects span innate immunity inflammation osteoclast function skeletal development and fertility CSF1R Protein Mouse (HEK293 Fc) is the recombinant mouse-derived CSF1R protein expressed by HEK293 with C-hFc labeled tag
Catalog No. 50-001-82696