Learn More
Waters Corp ACQUITY - XSelect CSH Phenyl-Hexyl Method Transfer Kit, 130Å, 1.7 - 5 µm, 2.1 - 4.6 mm X 50 - 150mm, 2/pk
Supplier:  Waters Corp 186005530
Encompass_Preferred
ACQUITY Method Transfer Kits contain an ACQUITY CSH column and matching XSelect HPLC column. The columns contained in an ACQUITY CSH Method Transfer Kit possess the same length to particle size (l/dp) ratio and will provide the same selectivity.
Catalog No. 50-791-487