Learn More
Cloud Clone Corp CSI032Ra01 Cloud-Clone Brand Primary Rat Subcutaneous Preadipocytes (SPrAD) Size 5X10^5 Ship with dry ice
Supplier:  Cloud Clone Corp CSI032RA01
Encompass_Preferred
Species: Rat. Tissue Source: Subcutaneous tissue. Growth Properties: Adherent. Cell Tpye: Preadipocytes. Research Field: Adipose cell series;Preadipocytes;Skeletal, articular and cutaneous systems;
Catalog No. NC3953284