Learn More
Sino Biological Peptide Substrates: CSKtide
Supplier:  Sino Biological C6358
Encompass_Preferred
The synthetic peptide CSKtide (KKKEEIYFFFG-NH2) contains a tyrosine protein kinase phosphorylation site and can be used for CSK and TNK1 assay.
Catalog No. 50-256-9502