Learn More
Medchemexpress LLC HY-112134 1mg Medchemexpress, CSN5i-3 CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1121341MG
Encompass_Preferred
Medchemexpress, HY-112134 1mg CSN5i-3 CAS: CSN5i-3 is a potent, selective and orally available inhibitor of CSN5, and inhibits CSN-catalysed Cul1 deneddylation with an IC50 value of 5.8 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9032