Learn More
Aviva Systems Biology Corp CSNK1A1L Antibody - middle region (ARP52809_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52809_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CSNK1A1L. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human CSNK1A1L. Alternate names: MGC33182, RP11-532O21.2, CK1ApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rat, Zebrafish
Catalog No. 50-285-770