Learn More
Medchemexpress LLC CSRM617 hydrochloride | 1353749-74-2 | 100.0% | 1 ML
SDP
Supplier:  Medchemexpress LLC HY122611A10MM 1ML
Encompass_Preferred
CSRM617 hydrochloride is a small-molecule inhibitor of the transcription factor ONECUT2 supplied as the hydrochloride salt. It is provided with high purity and is available as a 10 mM solution in DMSO or in multiple solid quantity options for research use.
  • Selective ONECUT2 inhibitor with reported binding in SPR assays.
  • High purity (99.97%).
  • Available as 10 mM solution in DMSO (1 ML) and as solid quantities (5-100 mg).
  • Molecular weight 291.69 g/mol; formula C10H14ClN3O5.
  • Suitable for biochemical assays and cell-based studies.
Catalog No. 50-002-30302