Learn More
Medchemexpress LLC CT-2584 | 166981-13-1 | 99.4% | 533.79 g/mol | C30H55N5O3 | 25 MG

Supplier: Medchemexpress LLC HY11898225MG
CT-2584 is a small-molecule anticancer research compound (CAS 166981-13-1) that reduces NKEF-B expression and induces mitochondrial reactive oxygen species (ROS), leading to tumor cell death. It is supplied as a white to off-white solid suitable for in vitro and in vivo pharmacology studies.
- Induces mitochondrial reactive oxygen species (ROS) to kill tumor cells.
- Reduces NKEF-B expression in tumor cell lines.
- High purity: 99.4%.
- Soluble in DMSO (100 mg/mL) and ethanol (5 mg/mL) with ultrasonic or heating assistance.
- Formulatable for in vivo use at ≥3 mg/mL in recommended vehicles.
- Stable as a powder at -20°C for up to 3 years.
- Molar mass 533.79 g/mol; formula C30H55N5O3.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.