Learn More
Selleck Chemical LLC CTB-E1107-25MG
SDP
Supplier:  Selleck Chemical LLC E110725MG
Encompass_Preferred
CTB is a p300 activator and the lowest docked energy for CTB and p300 is 7 72 kcal/mol
Catalog No. 50-313-5661