Learn More
Perkin Elmer US LLC CTFE Port Plug 1¼ -28
Supplier:  Perkin Elmer US LLC N8152718
Encompass_Preferred
This 1 -28 CTFE Port Plug is for use with NexION 1000/2000 ICP-MS series instruments. It is included in the NexION 1000/2000 SMARTintro Black Kit (N8150036).
Catalog No. 50-210-5488