Learn More
OriGene CTHRC1 CytoSection 25 slides
SDP
Supplier:  OriGene TS404348P5
Encompass_Preferred
Transient overexpression of CTHRC1 in HEK293T cells, FFPE control for IHC, ICC and ISH staining, 25 slides per pack
Catalog No. NC2944389