Learn More
CELLULAR TECHNOLOGY LTD CTL-CEF-002 CELL PEPTIDE
SDP
Supplier:  CELLULAR TECHNOLOGY LTD CTLCEF002
Encompass
CEF-MHC Class I Control Peptide Pool Plus. 32 HLa Class I-restricted T cell epitopes from Cytomegalovirus, Epstein-Barr virus, and Influenza virus.
Catalog No. NC9672163