Learn More
Genscript Corporation cTnT 26D7 mAb Mouse
Supplier:  Genscript Corporation V00402100
Encompass
Cardiac TnT (cTnT) is the largest subunit of the troponin complex which is composed of cTnT troponin I (TnI) troponin C (TnC) It is encoded by the TNNT2 gene with a calculated molecular weight of 34 6 kDa The cTnT is considered as a useful biomarker for the diagnosis of acute myocardial infarction
Catalog No. 50-000-58085
Only null left
Add to Cart
Request a Quote