Learn More
ABclonal Technology CTP synthase / CTPS Rabbit mAb
SDP
Supplier:  ABclonal Technology A2091220UL
Encompass_Preferred
This gene encodes an enzyme responsible for the catalytic conversion of UTP (uridine triphosphate) to CTP (cytidine triphospate) This reaction is an important step in the biosynthesis of phospholipids and nucleic acids Activity of this proten is important in the immune system and loss of function of this gene has been associated with immunodeficiency Alternative splicing results in multiple transcript variants
Catalog No. 50-000-07996