Learn More
Medchemexpress LLC CTP Synthetase-IN-1 | 2338811-71-3 | C20H19F3N6O3S2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1521495MG
Encompass_Preferred
CTP Synthetase-IN-1 is a potent, orally active cytidine 5'-triphosphate synthetase (CTPS) inhibitor with IC50s of 32 nM for human CTPS1 and 18 nM for human CTPS2. This compound also exhibits anti-inflammatory effects and is intended for research use only.
  • Potent, orally active CTPS inhibitor.
  • Targets human CTPS1 (IC50: 32 nM) and CTPS2 (IC50: 18 nM).
  • Exhibits anti-inflammatory effects.
  • Suitable for research use.
  • Shows significant pharmacological responses in mouse collagen-induced arthritis (CIA) model.
  • High purity (99.70%).
  • Soluble in DMSO (100 mg/mL).
Catalog No. 50-003-73583