Learn More
Abcam CTPB, p300 HAT activator, 1MG
Supplier:  Abcam AB1422241MG
Encompass_Preferred
MW 554.1 Da, Purity >99%. Small-molecule p300 histone acetyltransferase (HAT) activator. Benzamide. Active in vitro and in vivo.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6274