Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ctx-0294885 hydrochloride | 2319590-02-6 | 99.6% | 474.39 g/mol | C22H25Cl2N7O | 10 MG
SDP

Supplier:  Medchemexpress LLC HY15985A10MG

Encompass_Preferred

CTX-0294885 hydrochloride is a broad-spectrum kinase inhibitor used in biochemical and cellular research to profile kinase interactions and signaling pathways relevant to inflammation, diabetes, and cancer. It is supplied as a light yellow to yellow solid with high purity suitable for research applications.

  • Broad-spectrum kinase inhibition for kinome profiling and target discovery
  • High purity (99.6%) suitable for biochemical and cellular assays
  • Chemical formula C22H25Cl2N7O; molecular weight 474.39 g/mol
  • Light yellow to yellow solid appearance
  • Storage: 4°C sealed and dry; if in solvent, store at -80°C (6 months) or -20°C (1 month)
  • Supplied as a 10 MG research pack

Catalog No. 50-002-85449


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.