Learn More
Medchemexpress LLC HY-108473 5mg , CU-CPT 4a CAS:1279713-77-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1084735MG
Encompass_Preferred
HY-108473 5mg CU-CPT 4a CAS: 1279713-77-7 CU-CPT 4a (TLR3-IN-1) is a potent, highly selective TLR3 signaling inhibitor. CU-CPT 4a represses the expression of downstream signaling pathways mediated by the TLR3/dsRNA complex, including TNF-α and IL-1β. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1271