Learn More
Medchemexpress LLC CU-CPT 4a (TLR3-IN-1) | 1279713-77-7 | 99.5% | 377.82 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY10847350MG
Encompass_Preferred
CU-CPT 4a (TLR3-IN-1) is a potent, highly selective TLR3 signaling inhibitor. It represses the expression of downstream signaling pathways mediated by the TLR3/dsRNA complex, including TNF-α and IL-1β.
  • Potent, highly selective TLR3 signaling inhibitor.
  • Represses expression of downstream signaling pathways mediated by the TLR3/dsRNA complex (e.g., TNF-α and IL-1β).
  • IC50 of 3.44 μM for blocking Poly (I:C)-induced TLR3 activation in RAW 264.7 cells.
  • Competes with dsRNA for binding to TLR3 with a Ki of 2.96 μM.
  • For research use only.
Catalog No. 50-003-46951