Learn More
Selleck Chemical LLC CU-CPT-8m S4461-5mg
SDP
Supplier:  Selleck Chemical LLC S44615MG
Encompass_Preferred
CU-CPT-8m (CPD1578) is a specific antagonist of toll-like receptor 8(TLR8) with IC50 of 67 nM and Kd of 220 nM
Catalog No. 50-311-0207