Learn More
TARGETMOL CHEMICALS INC CU-CPT-8m 5MG
Supplier: TARGETMOL CHEMICALS INC T73475MG
Also available in 1 mL, 1 mg, 2 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. CU-CPT-8m (TLR8-specific antagonist) is an toll-like receptor 8 (TLR8) antagonist (IC50 67 nM)