Learn More
TARGETMOL CHEMICALS INC CU-CPT17e 1MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T108991MG
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes.CU-CPT17e is a potent agonist of TLR(TLR3, TLR8, and TLR9). Purity 99.18%
Catalog No. 50-287-8181