Learn More
Selleck Chemical LLC Cu-CPT22
SDP
Supplier:  Selleck Chemical LLC S867725MG
Encompass_Preferred
CU-CPT22 shows dose-dependent inhibitory effects blocking Pam3CSK4-induced TLR1/2 activation with an IC50 of 0 58 0 09 M while no significant inhibition to TLR2/6 It demonstrates minimal non-specific inhibition against a panel of 10 representative kinases (PDGFRB MET DDR2 SRC MAPK1 PAK1 AKT1 PKC- CAMK1 and PLK4)
Catalog No. 50-000-01233