Learn More
TARGETMOL CHEMICALS INC CU-CPT22 5MG
Supplier: TARGETMOL CHEMICALS INC T150205MG
Also available in 1 mL, 1 mg, 2 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. CU-CPT22 is the first probe for the complex between toll-like receptors TLR1 and TLR2. CU-CPT22 binds at the interface of TLR1 and TLR2 (IC50 = 0.58 uM). It competes with the synthetic triacylated lipoprotein (Pam3CSK4) binding to TLR1/2 (Ki 0.41 uM). Purity 99.3%