Learn More
Apexbio Technology LLC CU-CPT9b 2162962-69-6 5mg
SDP
Supplier:  Apexbio Technology LLC A47095
Encompass_Preferred
CU-CPT9b (CAS 2162962-69-6) is a selective antagonist of Toll-like receptor 8 (TLR8) designed to modulate immune signaling by inhibiting TLR8-mediated pathways The compound demonstrates high binding affinity to TLR8 with a reported Kd of 21 nM and an IC50 of 0 7 nM indicating potent inhibition of receptor activity CU-CPT9b is utilized in studies investigating immune regulation and inflammatory responses providing a valuable tool for exploring the role of TLR8 in the pathogenesis of immune and inflammation-related diseases and for the development of targeted therapeutic approaches
Catalog No. 50-000-93477