Learn More
Selleck Chemical LLC CU-T12-9 S2979-25mg
SDP
Supplier:  Selleck Chemical LLC S297925MG
Encompass_Preferred
CU-T12-9 is a specific agonist of Toll-like receptor (TLR) 1/2 with EC50 of 52 9 nM in HEK-Blue hTLR2 SEAP assay CU-T12-9 activates both the innate and the adaptive immune systems CU-T12-9 signals through NF- B and invokes an elevation of the downstream effectors TNF- IL-10 and iNOS
Catalog No. 50-311-1658