Learn More
Medchemexpress LLC Cuatsp | 68341-12-8 | 99.4% | 446.05 | C18H18CuN6S2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1398265MG
Encompass_Preferred
CuATSP is a copper-based small molecule used in research to inhibit ferroptotic cell death. It is reported to be approximately 20-fold more potent than related copper complexes and is supplied as a high-purity powder for laboratory studies of oxidative lipid peroxidation and cell death mechanisms. Store and handle under recommended laboratory conditions; not for clinical use.
Catalog No. 50-002-23063