Learn More
Medchemexpress LLC Cucurbitacin B | 6199-67-3 | 558.70 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYN04161MG
Encompass_Preferred
Cucurbitacin B is a highly oxidized tetracyclic triterpenoid that is orally active. It has been shown to inhibit tumor cell growth, migration, and invasion, induce cell apoptosis, and cause cell cycle arrest. Additionally, Cucurbitacin B possesses potent anti-inflammatory, antioxidant, antiviral, hypoglycemic, hepatoprotective, and neuroprotective activities.
  • Belongs to a class of highly oxidized tetracyclic triterpenoids
  • Oral active
  • Inhibits tumor cell growth
  • Inhibits tumor cell migration
  • Inhibits tumor cell invasion
  • Induces cell apoptosis
  • Induces cell cycle arrest
  • Potent anti-inflammatory activity
  • Potent antioxidant activity
  • Potent antiviral activity
  • Potent hypoglycemic activity
  • Potent hepatoprotective activity
  • Potent neuroprotective activity
Catalog No. 50-003-89921