Learn More
Cayman Chemical CucurbItacIn D 10mg
SDP
Supplier:  Cayman Chemical 3617210 MG
Encompass_Preferred
A triterpenoid; decreases levels of HER2, Raf, and Cdk6 in MCF-7 cell lysates; inhibits the proliferation of MCF-7 cells (IC50 = 0.598 µM); inhibits cigarette smoke condensate-induced HIV-1 replication in chronically HIV-1-infected U1 macrophages at 0.1 µM; decreases cold and mechanical allodynia in a mouse model of paclitaxel-induced neuropathic pain at 0.025 mg/kg
Catalog No. 50-291-1682