Learn More
Medchemexpress LLC Cucurbitacin D | 3877-86-9 | 516.67 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYN19861MG
Encompass_Preferred
Cucurbitacin D is the active ingredient in *Trichosanthes kirilowii*. It disrupts the interaction between Hsp90 and its co-chaperones, Cdc37 and p23. This compound acts as an inflammasome activator, inducing cell cycle arrest and apoptosis, and exhibits anti-tumor and anti-inflammatory effects.
  • Active ingredient derived from *Trichosanthes kirilowii*
  • Disrupts interaction between Hsp90 and its co-chaperones
  • Acts as an inflammasome activator
  • Induces cell cycle arrest
  • Induces cell apoptosis
  • Exhibits anti-tumor properties
  • Exhibits anti-inflammatory properties
Catalog No. 50-004-15240