Learn More
Cayman Chemical CucurbItacIn E 10mg
SDP
Supplier:  Cayman Chemical 1482110 MG
Encompass_Preferred
A natural triterpene; reduces MPP+-induced death of neuronal PC12 cells through inhibition of autophagy in vitro; inhibits growth of T24 bladder, MDA-MB-468 and MCF-7 breast, PC3 prostate, and colorectal cancer cell lines (IC50s = 50-1,000 nM) through induction of G2/M arrest and apoptosis; increases bilirubin binding to HSA in human plasma in a dose-dependent manner; inhibits depolymerization of actin filaments isolated from rabbit skeletal muscle actin and in HeLa cells
Catalog No. 50-296-9404