Learn More
Medchemexpress LLC HY-N0417 5mg Medchemexpress, Cucurbitacin E CAS:18444-66-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN04175MG
Encompass_Preferred
Medchemexpress, HY-N0417 5mg Cucurbitacin E CAS:18444-66-1 Cucurbitacin E is a natural compound which from the climbing stem of Cucumic melo L. Cucurbitacin E significantly suppresses the activity of the cyclin B1 / CDC2 complex. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3562