Learn More
Cayman Chemical CucurbItacIn I 1mg
SDP
Supplier:  Cayman Chemical 147471 MG
Encompass_Preferred
A potent inhibitor of the STAT3/JAK signaling pathway; suppresses levels of tyrosine phosphorylated STAT3, STAT3 DNA binding and STAT3-mediated gene transcription; suppresses tumor growth in vivo
Catalog No. 50-297-0773