Learn More
TARGETMOL CHEMICALS INC Verinurad 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T399425MG
Encompass_Preferred
Also available in 1 mL, 2 mg, 5 mg, 10 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Verinurad (RDEA3170) (RDEA3170) is an organic anion transporter URAT1 (SLC22A12) inhibitor for the treatment of gout and hyperuricemia. Purity 99.22%
Catalog No. 50-287-8817