Learn More
Medchemexpress LLC CUDA disodium | 98.0% | 386.48 | 10 MG
Supplier: Medchemexpress LLC HY121538A10MG
CUDA disodium is a potent inhibitor of soluble epoxide hydrolase (sEH), demonstrating IC50s of 11.1 nM for mouse sEH and 112 nM for human sEH. It selectively enhances peroxisome proliferator-activated receptor (PPAR) alpha activity. This compound is valuable for research into cardiovascular disease and is intended for research use only.
- Potent inhibitor of soluble epoxide hydrolase (sEH)
- Increases peroxisome proliferator-activated receptor (PPAR) alpha activity
- Functions as a PPAR alpha ligand
- Suitable for research in cardiovascular disease