Learn More
Medchemexpress LLC 7-[4-(3-ethynylanilino)-7-methoxyquinazolin-6-yl]oxy-N-hydroxyheptanamide | 1012054-59-9 | MFCD15528940 | >98.0% | 434.49 g/mol | C24H26N4O4 | 100 MG

Supplier: Medchemexpress LLC HY10223100MG
CUDC-101 is a small-molecule research compound that concurrently inhibits histone deacetylases (HDACs) and receptor tyrosine kinases EGFR and HER2. It is used in preclinical cancer research to study combined epigenetic and kinase-targeted therapies.
- Potent inhibition of HDAC, EGFR, and HER2 with IC50s of 4.4 nM, 2.4 nM, and 15.7 nM.
- Contains a hydroxamic acid moiety associated with HDAC activity.
- High purity (>98.0%) suitable for research applications.
- Soluble in DMSO at 25 mg/mL (approximately 57.54 mM; may require ultrasonic agitation).
- Molecular formula C24H26N4O4 and molecular weight 434.49 g/mol.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.