Learn More
Apexbio Technology LLC CUDC-907,5mg CAS# 1339928-25-4
SDP
Supplier:  Apexbio Technology LLC A40975
Encompass_Preferred
CUDC-907 is a dual-acting inhibitor of HDAC and PI3K with IC50 values of 1.7/5.0/1.8/2.8 nM (HDAC1/2/3/10) and 19/54/39 nM (PI3K).[1]The phosphoinositide 3-kinases (PI3Ks) contain three classes of PI3K each with its own distinct lipid products and specific substrate. They are a family of lipid kinases that regulates a wide range of pathway by propagating intracellular signaling cascades. PI3K phosphorylates the 3’-OH group of phosphatidylinositols. This activated AKT, the protein Ser/Thr-kinase, by recruiting them to the cell membrane. The PI3K/AKT signaling pathway is critical in cancer, because it promotes cell growth and survival. The studies have proved that PI3K pathway plays an important role in cancer progression and treatment for lung cancers, breast cancer.[2, 3] Other sizes are also available. Please inqury us for quote.
Catalog No. 50-224-0094