Learn More
Seca Corp seca 490 Cuff, Extension tube, 51 inches / 1.3 m length
SDP
Supplier:  Seca Corp 4900033001
Encompass_Preferred
The extension tube for cuffs of the seca 535 mVSA vital signs monitor simplifies your work and gives you more freedom of movement. • Air tube extension for seca 535 mVSA vital signs monitor cuffs • Tube length: 51 in / 1.3 m • Polyurethane (PU)
Catalog No. NC1834735